Assay Details
Target Gene Details
Entrez Gene ID: | 54584 |
Gene Name: | G protein subunit beta 1 like |
Gene Aliases: |
DGCRK3, FKSG1, GY2, WDR14, WDVCF |
Location: |
Chr.22:19788411-19854939 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNB1L | NM_053004.2 | NP_443730.1 | ||
AB051432.1 | ||||
AF238328.1 | AAG36826.1 | |||
AF301895.1 | AAG53933.1 | |||
AY007378.1 | AAG12162.1 | |||
BC012060.1 | AAH12060.1 | |||
CR456496.1 | CAG30382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834129 | Chr.22:19705617 - 19919068 on Build GRCh38 | Loss | C22orf29 GP1BB SEPT5-GP1BB SEPT5 TXNRD2 GNB1L TBX1 |
nsv517478 | Chr.22:19703979 - 19817740 on Build GRCh38 | Loss | GP1BB SEPT5-GP1BB SEPT5 GNB1L TBX1 |
esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain | LINC00895 MIR4761 RTN4R ARVCF MIR6816 TANGO2 GNB1L CCDC188 MIR1286 MIR3618 SEPT5-GP1BB LOC440792 SEPT5 ZDHHC8 TXNRD2 DGCR6L TRMT2A TBX1 MIR185 C22orf29 CDC45 LOC284865 COMT LOC101927859 LINC00896 DGCR8 GP1BB RANBP1 UFD1L CLDN5 MIR1306 |
nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss | MIR4761 RTN4R ARVCF MIR6816 TANGO2 GNB1L CCDC188 MIR1286 MIR3618 SEPT5-GP1BB LOC440792 SEPT5 ZDHHC8 TXNRD2 DGCR6L TRMT2A TBX1 MIR185 C22orf29 LOC284865 COMT LINC00896 DGCR8 GP1BB RANBP1 MIR1306 |
nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain | MIR4761 RTN4R ARVCF MIR6816 TANGO2 GNB1L CCDC188 MIR1286 MIR3618 SEPT5-GP1BB LOC440792 SEPT5 ZDHHC8 TXNRD2 DGCR6L TRMT2A TBX1 MIR185 C22orf29 LOC284865 COMT LINC00896 DGCR8 GP1BB RANBP1 MIR1306 |
nsv471181 | Chr.22:19729797 - 19815256 on Build GRCh38 | Loss | GNB1L TBX1 |
esv3568260 | Chr.22:19777375 - 19881883 on Build GRCh38 | Loss | C22orf29 TXNRD2 GNB1L TBX1 |
nsv438325 | Chr.22:19784656 - 19808063 on Build GRCh38 | Loss | GNB1L |
esv3647279 | Chr.22:19719848 - 19878980 on Build GRCh38 | Gain | C22orf29 GP1BB SEPT5-GP1BB SEPT5 TXNRD2 GNB1L TBX1 |
dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain | MIR185 C22orf29 MIR4761 RTN4R LOC284865 ARVCF COMT MIR6816 LINC00896 TANGO2 GNB1L CCDC188 MIR1286 MIR3618 DGCR8 LOC440792 RANBP1 ZDHHC8 TXNRD2 MIR1306 DGCR6L TRMT2A TBX1 |
esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain | C22orf39 LOC100996415 LINC00895 LOC100996401 TSSK2 MIR4761 SLC25A1 HIRA DGCR11 CLTCL1 PI4KAP1 DGCR6 MRPL40 GNB1L PRODH LINC01311 SEPT5-GP1BB USP18 SEPT5 RIMBP3 TXNRD2 LOC729461 TBX1 C22orf29 CDC45 LOC100996432 LOC102725072 DGCR5 DGCR2 GSC2 COMT DGCR9 GGT3P FAM230A DGCR10 DGCR14 GP1BB TMEM191B UFD1L CLDN5 LOC642643 |
nsv588216 | Chr.22:19086099 - 19839117 on Build GRCh38 | Gain | C22orf39 CDC45 LINC00895 TSSK2 SLC25A1 HIRA DGCR2 CLTCL1 GSC2 MRPL40 GNB1L LINC01311 DGCR14 GP1BB SEPT5-GP1BB SEPT5 UFD1L CLDN5 TBX1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |