Assay Details
Target Gene Details
Entrez Gene ID: | 8711 |
Gene Name: | tyrosine kinase non receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7380559-7389774 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNK1 | NM_001251902.1 | NP_001238831.1 | ||
NM_003985.4 | NP_003976.2 | |||
XM_011524045.2 | XP_011522347.1 | |||
XM_011524046.2 | XP_011522348.1 | |||
XM_011524047.2 | XP_011522349.1 | |||
XM_011524048.1 | XP_011522350.1 | |||
AK225403.1 | ||||
AK310684.1 | ||||
AK311063.1 | ||||
DA089305.1 | ||||
DB287365.1 | ||||
U43408.1 | AAC50427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | FXR2 POLR2A PLSCR3 SLC16A11 YBX2 BCL6B SENP3-EIF4A1 GABARAP NLGN2 LOC100996842 SENP3 SPEM1 SNORA48 MIR195 ALOX12 ALOX12-AS1 SAT2 EIF5A SOX15 ATP1B2 TMEM256-PLSCR3 KCTD11 PHF23 CTDNEP1 TMEM256 CLEC10A RNASEK SLC2A4 SHBG SLC35G6 MIR324 TMEM95 GPS2 NEURL4 MIR497HG TNFSF12 TNK1 ACAP1 ELP5 ACADVL C17orf74 ZBTB4 CD68 DLG4 CLDN7 TNFSF13 DVL2 MPDU1 MIR497 C17orf49 RNASEK-C17orf49 SLC16A13 ALOX12P2 ASGR2 EIF4A1 CHRNB1 TMEM102 TNFSF12-TNFSF13 LOC107983988 FBXO39 SNORA67 ASGR1 FGF11 SNORD10 TEKT1 |
nsv523672 | Chr.17:7379825 - 7549660 on Build GRCh38 | Loss | POLR2A TNFSF12 TNK1 PLSCR3 C17orf74 CHRNB1 ZBTB4 TMEM102 TNFSF12-TNFSF13 TMEM256-PLSCR3 NLGN2 LOC107983988 TMEM256 FGF11 SLC35G6 SPEM1 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | NEURL4 TNK1 PLSCR3 ACAP1 ELP5 ACADVL C17orf74 YBX2 GABARAP NLGN2 DLG4 CLDN7 DVL2 SPEM1 EIF5A CHRNB1 TMEM102 TMEM256-PLSCR3 KCTD11 PHF23 CTDNEP1 LOC107983988 TMEM256 ASGR1 FGF11 SLC2A4 MIR324 TMEM95 GPS2 |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | FXR2 POLR2A PLSCR3 SLC16A11 TP53 YBX2 BCL6B SENP3-EIF4A1 GABARAP NLGN2 LOC100996842 SENP3 SPEM1 SNORA48 MIR195 ALOX12 ALOX12-AS1 SAT2 EIF5A SOX15 ATP1B2 TMEM256-PLSCR3 KCTD11 PHF23 CTDNEP1 WRAP53 TMEM256 CLEC10A RNASEK SLC2A4 SHBG SLC35G6 MIR324 TMEM95 GPS2 NEURL4 MIR497HG TNFSF12 TNK1 ACAP1 ELP5 ACADVL C17orf74 ZBTB4 CD68 DLG4 CLDN7 TNFSF13 DVL2 MPDU1 MIR497 C17orf49 RNASEK-C17orf49 SLC16A13 ASGR2 EIF4A1 CHRNB1 TMEM102 TNFSF12-TNFSF13 LOC107983988 SNORA67 ASGR1 FGF11 SNORD10 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |