Assay Details
Target Gene Details
Entrez Gene ID: | 338707 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyltransferase 4 |
Gene Aliases: |
- |
Location: |
Chr.11:369773-382117 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
B4GALNT4 | NM_178537.4 | 14 | 1864 | NP_848632.2 |
XM_017017654.1 | 14 | 3176 | XP_016873143.1 | |
AB089939.1 | 14 | 1855 | BAD06471.1 | |
BC131528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527327 | Chr.11:295343 - 620599 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv552776 | Chr.11:366710 - 432436 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv1118197 | Chr.11:374500 - 377100 on Build GRCh38 | Deletion |
![]() |
esv28392 | Chr.11:363975 - 403432 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv951268 | Chr.11:368601 - 503700 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv552770 | Chr.11:356090 - 444628 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv1505n54 | Chr.11:198510 - 470331 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1159790 | Chr.11:340219 - 491334 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv1507n54 | Chr.11:368778 - 382912 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)