Assay Details
Target Gene Details
Entrez Gene ID: | 2954 |
Gene Name: | glutathione S-transferase zeta 1 |
Gene Aliases: |
GSTZ1-1, MAAI, MAI |
Location: |
Chr.14:77320887-77331597 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTZ1 | NM_001312660.1 | 1 | 155 | NP_001299589.1 |
NM_145870.2 | 1 | 155 | NP_665877.1 | |
NM_145871.2 | 1 | 155 | NP_665878.2 | |
AJ001838.1 | CAA05045.1 | |||
AK309054.1 | ||||
AK315154.1 | 1 | 46 | ||
AK315609.1 | ||||
BC001453.1 | 1 | 31 | AAH01453.1 | |
BG330365.1 | ||||
BG772450.1 | 1 | 162 | ||
U86529.1 | AAB96392.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |