Assay Details
Target Gene Details
Entrez Gene ID: | 9079 |
Gene Name: | LIM domain binding 2 |
Gene Aliases: |
CLIM1, LDB-2, LDB1 |
Location: |
Chr.4:16501534-16898809 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527861 | Chr.4:15848363 - 16941367 on Build GRCh38 | Gain | LDB2 CD38 TAPT1-AS1 TAPT1 FGFBP1 PROM1 FGFBP2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |