Assay Details
Target Gene Details
Entrez Gene ID: | 51528 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
C14orf100, CDA06, HSPC213, HSPC327, JAMP |
Location: |
Chr.14:59484443-59505407 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JKAMP | NM_001098625.2 | 3 | 279 | NP_001092095.1 |
NM_001284201.1 | 3 | 820 | NP_001271130.1 | |
NM_001284202.1 | 4 | 406 | NP_001271131.1 | |
NM_001284203.1 | 3 | 329 | NP_001271132.1 | |
NM_001284204.1 | 3 | 297 | NP_001271133.1 | |
NM_016475.4 | 3 | 297 | NP_057559.2 | |
XM_006720171.3 | 3 | 273 | XP_006720234.1 | |
XM_017021366.1 | 3 | 206 | XP_016876855.1 | |
AF151047.1 | 3 | 219 | AAF36133.1 | |
AF161445.1 | 3 | 218 | AAF29005.1 | |
AF212245.1 | 3 | 219 | AAG41781.1 | |
AF529304.1 | 1 | 534 | AAP85636.1 | |
AK021683.1 | 3 | 178 | BAB13874.1 | |
AK027591.1 | 3 | 223 | BAB55216.1 | |
AK056145.1 | 3 | 696 | ||
AK298209.1 | 3 | 204 | ||
AL136696.1 | 3 | 189 | CAB66631.1 | |
BC005198.1 | 3 | 205 | AAH05198.1 | |
BC010359.1 | 3 | 236 | AAH10359.1 | |
BU928810.1 | 3 | 226 | ||
BX161501.1 | 4 | 289 | CAD61945.1 | |
CR457163.1 | 2 | 132 | CAG33444.1 | |
CR533510.1 | 2 | 150 | CAG38541.1 | |
DC381037.1 | 3 | 297 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |