Assay Details
Target Gene Details
Entrez Gene ID: | 10419 |
Gene Name: | protein arginine methyltransferase 5 |
Gene Aliases: |
HRMT1L5, IBP72, JBP1, SKB1, SKB1Hs |
Location: |
Chr.14:22920511-22929585 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT5 | NM_001039619.2 | NP_001034708.1 | ||
NM_001282953.1 | NP_001269882.1 | |||
NM_001282954.1 | NP_001269883.1 | |||
NM_001282955.1 | NP_001269884.1 | |||
NM_001282956.1 | NP_001269885.1 | |||
NM_006109.4 | NP_006100.2 | |||
XM_017020927.1 | XP_016876416.1 | |||
AB451246.1 | ||||
AB451370.1 | ||||
AF015913.1 | AAB66581.1 | |||
AF167572.1 | AAF04502.1 | |||
AK075251.1 | ||||
AK300863.1 | ||||
AK301812.1 | ||||
AK302240.1 | ||||
AK315500.1 | ||||
AL833612.1 | ||||
BC025979.2 | AAH25979.1 | |||
CR456741.1 | CAG33022.1 |
Target Gene Details
Entrez Gene ID: | 100505758 |
Gene Name: | PRMT5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.14:22919456-22923407 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRMT5-AS1 | NR_120599.1 | 1 | 1576 | |
NR_120600.1 | 1 | 1576 | ||
AK093277.1 | 1 | 1576 | ||
DB548852.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |