Assay Details
Target Gene Details
Entrez Gene ID: | 124056 |
Gene Name: | NADPH oxidase organizer 1 |
Gene Aliases: |
P41NOX, P41NOXA, P41NOXB, P41NOXC, SH3PXD5, SNX28 |
Location: |
Chr.16:1978917-1982751 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOXO1 | NM_001267721.1 | NP_001254650.1 | ||
NM_144603.3 | NP_653204.1 | |||
NM_172167.2 | NP_751907.1 | |||
NM_172168.2 | NP_751908.1 | |||
XM_005255099.4 | XP_005255156.1 | |||
XM_017022927.1 | XP_016878416.1 | |||
XM_017022928.1 | XP_016878417.1 | |||
AB097667.1 | BAC76711.1 | |||
AF532983.1 | AAM97925.1 | |||
AF532984.1 | AAM97926.1 | |||
AF532985.1 | AAM97927.1 | |||
AF539796.1 | AAN75141.1 | |||
AK292197.1 | ||||
AK310198.1 | ||||
AK313917.1 | ||||
AY191359.1 | AAO38665.1 | |||
AY255768.1 | AAP13479.1 | |||
BC015917.2 | AAH15917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv521569 | Chr.16:1971892 - 2035996 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4858n54 | Chr.16:1955562 - 2038584 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952901 | Chr.16:1888800 - 2135899 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160329 | Chr.16:1961727 - 2128574 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv4860n54 | Chr.16:1974897 - 1980689 on Build GRCh38 | Loss |
![]() ![]() |
nsv471070 | Chr.16:1947613 - 2024218 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833120 | Chr.16:1893102 - 1980660 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv336n27 | Chr.16:1965120 - 2038584 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)