Assay Details
Target Gene Details
Entrez Gene ID: | 1655 |
Gene Name: | DEAD-box helicase 5 |
Gene Aliases: |
G17P1, HLR1, HUMP68, p68 |
Location: |
Chr.17:64498254-64507038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX5 | NM_001320595.1 | NP_001307524.1 | ||
NM_001320596.1 | NP_001307525.1 | |||
NM_001320597.1 | NP_001307526.1 | |||
NM_004396.4 | NP_004387.1 | |||
AB209919.1 | 11 | 2600 | BAD93156.1 | |
AB451257.1 | ||||
AB451382.1 | ||||
AK055995.1 | 1 | 710 | ||
AK297192.1 | ||||
AK297753.1 | ||||
AK297905.1 | ||||
AK310318.1 | ||||
BC016027.1 | AAH16027.1 | |||
BT006943.1 | AAP35589.1 | |||
BX571764.1 | 11 | 2617 | CAE11890.1 | |
X15729.1 | CAA33751.1 | |||
X52104.1 | CAA36324.1 |
Target Gene Details
Entrez Gene ID: | 100616387 |
Gene Name: | microRNA 3064 |
Gene Aliases: |
- |
Location: |
Chr.17:64500774-64500839 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIR3064 | NR_039891.2 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |