Assay Details
Target Gene Details
Entrez Gene ID: | 23179 |
Gene Name: | ral guanine nucleotide dissociation stimulator like 1 |
Gene Aliases: |
RGL |
Location: |
Chr.1:183636047-183928551 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGL1 | NM_001297669.1 | 1 | 120 | NP_001284598.1 |
NM_001297670.1 | 1 | 120 | NP_001284599.1 | |
NM_015149.4 | 1 | 120 | NP_055964.3 | |
XM_011509339.2 | 1 | 56 | XP_011507641.1 | |
XM_011509341.1 | 1 | 73 | XP_011507643.1 | |
XM_011509342.2 | 1 | 73 | XP_011507644.1 | |
AF186780.1 | 1 | 82 | AAF67281.1 | |
AK295179.1 | ||||
AK316526.1 | 1 | 58 | ||
BP235344.1 | 1 | 94 | ||
BX537416.1 | 1 | 60 | CAD97658.1 | |
DC333918.1 | 1 | 120 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |