Assay Details
Target Gene Details
Entrez Gene ID: | 10367 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
CALC, CBARA1, EFHA3, MPXPS |
Location: |
Chr.10:72367326-72626191 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MICU1 | NM_001195518.1 | NP_001182447.1 | ||
NM_001195519.1 | NP_001182448.1 | |||
NM_006077.3 | NP_006068.2 | |||
XM_005269383.2 | XP_005269440.1 | |||
XM_005269386.2 | XP_005269443.1 | |||
XM_011539119.1 | XP_011537421.1 | |||
AK022697.1 | BAB14187.1 | |||
AK023318.1 | ||||
AK225298.1 | ||||
AK225695.1 | ||||
AK296086.1 | ||||
AK298347.1 | ||||
AL117423.1 | CAB55915.1 | |||
BC004190.2 | AAH04190.1 | |||
BC004216.2 | AAH04216.1 | |||
BC016641.1 | AAH16641.1 | |||
Y17711.1 | CAA76830.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |