Assay Details
Target Gene Details
Entrez Gene ID: | 283337 |
Gene Name: | zinc finger protein 740 |
Gene Aliases: |
Zfp740 |
Location: |
Chr.12:53180715-53190870 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF740 | NM_001004304.3 | 7 | 1561 | NP_001004304.1 |
XM_006719344.2 | XP_006719407.1 | |||
XM_011538194.2 | XP_011536496.1 | |||
XM_017019181.1 | 7 | 1795 | XP_016874670.1 | |
AK091946.1 | 4 | 944 | ||
BC053557.1 | 7 | 1564 | AAH53557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | CSAD SOAT2 SPRYD3 ZNF740 RARG IGFBP6 MFSD5 ITGB7 |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | CSAD SOAT2 ESPL1 ZNF740 RARG MFSD5 ITGB7 |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | CSAD SOAT2 ZNF740 RARG IGFBP6 ITGB7 |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | CSAD EIF4B SPRYD3 ESPL1 TNS2 MIR6757 LOC283335 ZNF740 RARG ITGB7 SOAT2 PFDN5 SP7 C12orf10 IGFBP6 MFSD5 AAAS |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | CSAD SOAT2 ZNF740 RARG ITGB7 |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | CSAD SOAT2 SPRYD3 TNS2 ZNF740 RARG IGFBP6 ITGB7 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |