Assay Details
Target Gene Details
Entrez Gene ID: | 60526 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
C2orf43, hLDAH |
Location: |
Chr.2:20682489-20823139 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LDAH | NM_001282719.1 | 6 | 2138 | NP_001269648.1 |
NM_001282720.1 | 6 | 2120 | NP_001269649.1 | |
NM_001282721.1 | 6 | 2108 | NP_001269650.1 | |
NM_001282722.1 | 5 | 1964 | NP_001269651.1 | |
NM_001282723.1 | 6 | 2029 | NP_001269652.1 | |
NM_001282724.1 | 5 | 1903 | NP_001269653.1 | |
NM_021925.3 | 7 | 2264 | NP_068744.1 | |
NR_104233.1 | 5 | 1873 | ||
XM_011533014.2 | 7 | 2291 | XP_011531316.1 | |
AK021672.1 | 3 | 1486 | ||
AK025473.1 | 7 | 2202 | BAB15142.1 | |
AL137397.1 | 1 | 786 | ||
BC017473.1 | 7 | 2221 | AAH17473.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |