Assay Details
Target Gene Details
Entrez Gene ID: | 55101 |
Gene Name: | ATP5S like |
Gene Aliases: |
- |
Location: |
Chr.19:41431318-41447822 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP5SL | NM_001167867.1 | 6 | 1152 | NP_001161339.1 |
NM_001167868.1 | 5 | 989 | NP_001161340.1 | |
NM_001167869.1 | 5 | 943 | NP_001161341.1 | |
NM_001167870.1 | 4 | 862 | NP_001161342.1 | |
NM_001167871.1 | 5 | 1025 | NP_001161343.1 | |
NM_001320838.1 | 5 | 906 | NP_001307767.1 | |
NM_001320840.1 | 5 | 1071 | NP_001307769.1 | |
NM_001320841.1 | 4 | 908 | NP_001307770.1 | |
NM_018035.2 | 6 | 1106 | NP_060505.2 | |
NR_030765.1 | 6 | 1089 | ||
NR_135476.1 | 5 | 957 | ||
NR_135477.1 | 6 | 1103 | ||
NR_135478.1 | 6 | 1120 | ||
XM_011527065.2 | 6 | 1157 | XP_011525367.1 | |
AK001103.1 | 6 | 1073 | BAA91503.1 | |
AK123631.1 | 3 | 1698 | ||
AK293132.1 | 5 | 952 | ||
AK297698.1 | 5 | 913 | ||
AK298197.1 | 5 | 1024 | ||
BC013323.1 | 4 | 838 | AAH13323.1 |