Assay Details
Target Gene Details
Entrez Gene ID: | 6405 |
Gene Name: | semaphorin 3F |
Gene Aliases: |
SEMA-IV, SEMA4, SEMAK |
Location: |
Chr.3:50155045-50189075 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SEMA3F | NM_001318798.1 | NP_001305727.1 | ||
NM_001318800.1 | NP_001305729.1 | |||
NM_004186.4 | NP_004177.3 | |||
XM_005265381.4 | XP_005265438.1 | |||
XM_005265382.4 | XP_005265439.1 | |||
XM_006713289.3 | XP_006713352.1 | |||
XM_006713290.3 | XP_006713353.1 | |||
XM_011533998.2 | XP_011532300.1 | |||
XM_011534000.2 | XP_011532302.1 | |||
AB209259.1 | BAD92496.1 | |||
BC042914.1 | AAH42914.1 | |||
U33920.1 | AAC50568.1 | |||
U38276.1 | AAB18276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759148 | Chr.3:50161045 - 50333946 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523560 | Chr.3:50134964 - 50314769 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv602n27 | Chr.3:50140578 - 50287241 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834692 | Chr.3:50161052 - 50356033 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv8316n54 | Chr.3:50115923 - 50299991 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv515986 | Chr.3:50066874 - 50254353 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470561 | Chr.3:50121341 - 50547568 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523662 | Chr.3:50107518 - 50547568 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv834690 | Chr.3:50064402 - 50263192 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv954855 | Chr.3:50152468 - 50439169 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)