Assay Details
Target Gene Details
Entrez Gene ID: | 2208 |
Gene Name: | Fc fragment of IgE receptor II |
Gene Aliases: |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF |
Location: |
Chr.19:7688757-7702755 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER2 | NM_001207019.2 | NP_001193948.2 | ||
NM_001220500.1 | NP_001207429.1 | |||
NM_002002.4 | NP_001993.2 | |||
XM_005272462.4 | XP_005272519.1 | |||
AK223207.1 | BAD96927.1 | |||
AK310071.1 | ||||
AK310707.1 | ||||
BC014108.2 | AAH14108.1 | |||
BC062591.1 | AAH62591.1 | |||
DA957269.1 | 3 | 328 | ||
M14766.1 | AAA52435.1 | |||
M15059.1 | AAA52434.1 | |||
X04772.1 | CAA28465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953959 | Chr.19:7669315 - 7700414 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)