Assay Details
Target Gene Details
Entrez Gene ID: | 55022 |
Gene Name: | phosphotyrosine interaction domain containing 1 |
Gene Aliases: |
HMFN2073, NYGGF4, P-CLI1, PCLI1 |
Location: |
Chr.2:229023973-229271341 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PID1 | NM_001100818.1 | 3 | 595 | NP_001094288.1 |
NM_017933.4 | 4 | 806 | NP_060403.3 | |
XM_011511384.2 | 3 | 478 | XP_011509686.1 | |
XM_017004404.1 | 4 | 5737 | XP_016859893.1 | |
XM_017004405.1 | 5 | 676 | XP_016859894.1 | |
XM_017004406.1 | 3 | 334 | XP_016859895.1 | |
XM_017004407.1 | 2 | 391 | XP_016859896.1 | |
AB075874.1 | 4 | 461 | BAD38656.1 | |
AK000708.1 | 3 | 581 | BAA91333.1 | |
AK096636.1 | 3 | 537 | ||
AK125359.1 | 4 | 730 | BAC86145.1 | |
AY317148.1 | 3 | 427 | AAP79437.1 | |
BC040164.1 | 2 | 382 | AAH40164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3195 | Chr.2:228989304 - 229033824 on Build GRCh38 | Deletion | PID1 |
nsv834563 | Chr.2:228864427 - 229064544 on Build GRCh38 | Gain | PID1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |