Assay Details
Target Gene Details
Entrez Gene ID: | 16 |
Gene Name: | alanyl-tRNA synthetase |
Gene Aliases: |
CMT2N, EIEE29 |
Location: |
Chr.16:70252295-70289552 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AARS | NM_001605.2 | NP_001596.2 | ||
AK222824.1 | BAD96544.1 | |||
AK225116.1 | ||||
AK299098.1 | ||||
BC011451.1 | AAH11451.1 | |||
BU178772.1 | ||||
D32050.1 | BAA06808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33745 | Chr.16:70082696 - 70285970 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3638952 | Chr.16:70252544 - 70262994 on Build GRCh38 | Gain |
![]() |
esv2758652 | Chr.16:69821219 - 70279319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1061286 | Chr.16:70219906 - 70297504 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv428328 | Chr.16:69952660 - 70279319 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833271 | Chr.16:70151791 - 70319431 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)