Assay Details
Target Gene Details
Entrez Gene ID: | 79866 |
Gene Name: | bora, aurora kinase A activator |
Gene Aliases: |
C13orf34 |
Location: |
Chr.13:72727739-72756198 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BORA | NM_001286746.1 | 12 | 2367 | NP_001273675.1 |
NM_001286747.1 | 11 | 2162 | NP_001273676.1 | |
NM_024808.3 | 12 | 2269 | NP_079084.3 | |
AK291496.1 | ||||
BC007119.1 | 3 | 1053 | AAH07119.1 | |
BC025367.2 | 12 | 2080 | AAH25367.2 | |
BC044213.1 | 11 | 1893 | AAH44213.1 |
Target Gene Details
Entrez Gene ID: | 22894 |
Gene Name: | DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease |
Gene Aliases: |
2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p |
Location: |
Chr.13:72755402-72782128 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DIS3 | NM_001128226.2 | 21 | 7347 | NP_001121698.1 |
NM_001322348.1 | 20 | 7279 | NP_001309277.1 | |
NM_001322349.1 | 22 | 7505 | NP_001309278.1 | |
NM_014953.4 | 21 | 7437 | NP_055768.3 | |
AF035310.1 | 1 | 1682 | ||
AF330044.1 | 21 | 7177 | AAL37479.1 | |
U79246.1 | 1 | 1682 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1049859 | Chr.13:72741128 - 72872273 on Build GRCh38 | Loss | DIS3 PIBF1 BORA |
More Information
Additional Information:
For this assay, SNP(s) [rs75805370] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic DGV Variation |