Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | 2 | 1338 | NP_005348.2 |
XM_005258937.3 | 2 | 1333 | XP_005258994.1 | |
XM_005258938.4 | 2 | 324 | XP_005258995.1 | |
XM_005258939.3 | 3 | 362 | XP_005258996.2 | |
XM_005258940.3 | 2 | 263 | XP_005258997.1 | |
XM_006723218.2 | 2 | 463 | XP_006723281.1 | |
XM_017026810.1 | 2 | 203 | XP_016882299.1 | |
AK292482.1 | 2 | 1318 | ||
BC070041.1 | 2 | 1338 | AAH70041.1 | |
U40002.1 | 2 | 1338 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | CNFN LOC101930071 LIPE LIPE-AS1 |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | CIC POU2F2 GRIK5 CD79A ARHGEF1 LOC100505622 MEGF8 MIR8077 RABAC1 ERF PAFAH1B3 LOC100505585 DEDD2 RPS19 CNFN LOC101930071 TMEM145 LIPE-AS1 LOC390937 MIR4323 PRR19 LIPE ZNF526 ATP1A3 GSK3A MIR6797 ZNF574 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | CIC POU2F2 CXCL17 CNFN LOC101930071 TMEM145 LIPE-AS1 CEACAM1 MEGF8 MIR8077 PRR19 ERF LIPE ZNF526 GSK3A PAFAH1B3 DEDD2 CEACAM8 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CIC POU2F2 GRIK5 CD79A CEACAM3 ARHGEF1 CEACAM7 LOC100505622 MEGF8 MIR8077 RABAC1 CEACAM4 ERF PAFAH1B3 LOC100505585 DEDD2 RPS19 CNFN LYPD4 LOC101930071 TMEM145 CEACAM5 LIPE-AS1 LOC390937 CEACAM6 MIR4323 PRR19 LIPE ZNF526 ATP1A3 GSK3A MIR6797 DMRTC2 ZNF574 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | CIC CXCL17 PSG8 CNFN LOC101930071 PSG11 PSG1 TMEM145 PSG7 PSG3 LIPE-AS1 LOC100289650 CEACAM1 PSG2 MEGF8 MIR8077 PSG10P PRR19 ERF LIPE GSK3A PAFAH1B3 PSG6 CEACAM8 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |