Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | NP_001139528.1 | ||
NM_001146082.2 | NP_001139554.1 | |||
NM_001146083.2 | NP_001139555.1 | |||
NM_024298.4 | NP_077274.3 | |||
XM_011527299.2 | XP_011525601.1 | |||
XM_011527300.2 | XP_011525602.1 | |||
XM_017027296.1 | XP_016882785.1 | |||
AK055908.1 | BAB71043.1 | |||
AK122789.1 | ||||
AK298689.1 | ||||
AK311493.1 | ||||
BC002512.2 | AAH02512.2 | |||
BC003164.1 | AAH03164.2 | |||
BC006309.2 | AAH06309.1 | |||
BC015857.1 | ||||
DA338950.1 | ||||
EU016381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1066425 | Chr.19:54182441 - 54237431 on Build GRCh38 | Gain | RPS9 TSEN34 LILRA6 MBOAT7 LILRB3 |
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | TMC4 LENG1 TSEN34 MBOAT7 |
nsv2546 | Chr.19:54182614 - 54270671 on Build GRCh38 | Deletion | RPS9 TSEN34 LILRA6 MBOAT7 LILRB5 LILRB3 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | PRPF31 OSCAR TMC4 LENG1 TSEN34 CNOT3 NDUFA3 TFPT MBOAT7 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR518C MIR1283-1 ZNF761 MIR520E ZNF677 ZNF813 LILRB2 MIR512-2 MIR520A MIR519D ZNF845 PRPF31 MYADM MIR515-2 TARM1 LENG1 MIR517C ZNF665 LILRB5 LOC105372457 MIR526B MIR518E MIR518F MIR372 MIR516A2 CACNG7 MIR371A MIR518B MIR519C CACNG8 ZNF331 FAM90A27P MIR4752 VSTM1 DPRX MIR520B MIR515-1 ZNF818P OSCAR MIR519E MIR935 MIR516A1 MIR522 MIR520G MIR526A2 MIR498 MIR518A2 MIR373 RPS9 LILRA5 TSEN34 MIR519A2 TFPT TPM3P9 MIR527 PRKCG BIRC8 MIR518A1 VN1R2 MIR519A1 TMC4 MIR516B1 CNOT3 CACNG6 MIR517A LILRA6 MIR371B MIR516B2 MIR525 ZNF525 MIR524 NDUFA3 MBOAT7 MIR1323 ZNF765 MIR1283-2 MIR521-1 MIR517B MIR520F MIR521-2 MIR523 MIR520H MIR520D MIR518D MIR520C MIR512-1 MIR526A1 MIR519B NLRP12 LILRB3 VN1R4 LOC284379 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | TMC4 LENG1 RPS9 TSEN34 LILRA6 MBOAT7 LILRB5 LILRB3 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LENG8 NCR1 LAIR1 KIR2DL4 RPS9 LILRA5 TSEN34 LILRB2 KIR3DL3 KIR3DX1 LENG8-AS1 LENG9 LILRB4 LILRB1 LILRP2 LAIR2 LOC101928804 TMC4 NLRP7 LILRA6 LILRB5 KIR3DL2 CDC42EP5 LILRA4 KIR2DS4 FCAR MIR4752 MBOAT7 LILRA1 TTYH1 MIR8061 KIR3DL1 KIR2DL1 LOC102724300 LILRA2 KIR2DL3 LILRB3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |