Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011671.1 | XP_016867160.1 | |||
XM_017011672.1 | XP_016867161.1 | |||
AA926779.1 | ||||
AB010447.1 | BAA84579.1 | |||
AB016542.1 | BAA36704.1 | |||
AF096296.1 | AAF09265.1 | |||
AF124601.1 | AAD22197.1 | |||
AF142434.1 | AAD46495.1 | |||
AY358893.1 | AAQ89252.1 | |||
BC069394.1 | AAH69394.1 | |||
BC093773.1 | AAH93773.1 | |||
BC101665.1 | AAI01666.1 | |||
BG211121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | CCL26 HIP1 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | MDH2 FDPSP2 GTF2IP1 STYXL1 SPDYE16 RHBDD2 STAG3L1 SNORA14A SRRM3 SSC4D LOC100133091 PMS2P2 NSUN5P1 GTF2IRD2B SPDYE5 LOC105375355 PMS2P10 MIR4651 TMEM120A YWHAG GTF2IP7 POM121C CCL26 NCF1C ZP3 CCL24 PMS2P3 HSPB1 DTX2 TRIM73 LOC441259 LOC107986711 HIP1 LOC541473 POMZP3 POR |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL26 HIP1 CCL24 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | CCL26 HIP1 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL26 HIP1 CCL24 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | MDH2 FDPSP2 STYXL1 SPDYE16 RHBDD2 SNORA14A SRRM3 SSC4D LOC100133091 SPDYE5 LOC105375355 MIR4651 TMEM120A YWHAG GTF2IP7 POM121C CCL26 ZP3 CCL24 PMS2P3 HSPB1 DTX2 LOC107986711 HIP1 POMZP3 POR |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |