Assay Details
Target Gene Details
Entrez Gene ID: | 79924 |
Gene Name: | adrenomedullin 2 |
Gene Aliases: |
AM2, dJ579N16.4 |
Location: |
Chr.22:50481724-50486440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADM2 | NM_001253845.1 | 3 | 1059 | NP_001240774.1 |
XM_017028940.1 | 2 | 1077 | XP_016884429.1 | |
AK024788.1 | 2 | 1300 | BAB15004.1 | |
AK090635.1 | 2 | 1077 | ||
AK225097.1 | 2 | 1300 | ||
BX400262.2 | ||||
CR456479.1 | CAG30365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3669 | Chr.22:50475511 - 50509415 on Build GRCh38 | Insertion | ADM2 NCAPH2 MIOX LMF2 |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion | SBF1 ADM2 SYCE3 NCAPH2 CHKB-CPT1B MIOX KLHDC7B TYMP LMF2 SCO2 ODF3B CPT1B |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain | SYCE3 NCAPH2 CHKB-CPT1B MIOX KLHDC7B SCO2 CHKB CPT1B CHKB-AS1 ADM2 TYMP LMF2 ODF3B |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss | LOC107985556 SBF1 ADM2 SYCE3 NCAPH2 MIOX KLHDC7B PPP6R2 TYMP LMF2 SCO2 ODF3B |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss | SBF1 SYCE3 NCAPH2 CHKB-CPT1B MIOX KLHDC7B SCO2 CHKB CPT1B ADM2 PPP6R2 TYMP LMF2 ODF3B |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion | LOC107985556 SBF1 ADM2 NCAPH2 MIOX PPP6R2 TYMP LMF2 SCO2 ODF3B |
nsv829338 | Chr.22:50440870 - 50489635 on Build GRCh38 | Gain | SBF1 ADM2 MIOX PPP6R2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |