Assay Details
Target Gene Details
Entrez Gene ID: | 9319 |
Gene Name: | thyroid hormone receptor interactor 13 |
Gene Aliases: |
16E1BP |
Location: |
Chr.5:892854-919348 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TRIP13 | NM_001166260.1 | NP_001159732.1 | ||
NM_004237.3 | NP_004228.1 | |||
XM_011514163.2 | XP_011512465.1 | |||
AK310376.1 | ||||
BC000404.2 | AAH00404.1 | |||
BC019294.1 | AAH19294.1 | |||
BG722761.1 | ||||
CR456744.1 | CAG33025.1 | |||
L40384.1 | AAC41732.1 | |||
U96131.1 | AAB64095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830186 | Chr.5:845005 - 966855 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv198e55 | Chr.5:685389 - 919382 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv2759316 | Chr.5:620048 - 1161361 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv596629 | Chr.5:457146 - 1468172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv10654 | Chr.5:676170 - 1082573 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509043 | Chr.5:747066 - 915204 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)