Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | KIF5A DTX3 INHBE MARS INHBC MBD6 GLI1 DDIT3 MIR616 MIR6758 PIP4K2C DCTN2 ARHGAP9 |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss | KIF5A DTX3 ARHGEF25 MARS LOC101927583 MBD6 GLI1 DDIT3 MIR616 MIR6758 PIP4K2C DCTN2 ARHGAP9 |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | INHBC MBD6 GLI1 DDIT3 MIR616 MIR6758 INHBE ARHGAP9 MARS |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | SNORD59A CNPY2 RDH16 MYO1A MARS SHMT2 STAC3 RNF41 STAT6 MIR616 ATP5B NAB2 LRP1-AS KIF5A NACA PAN2 LOC101927583 MBD6 NXPH4 IL23A HBCBP MIP B4GALNT1 APOF PRIM1 NEMP1 SLC26A10 SLC39A5 SDR9C7 ARHGEF25 INHBE CS NDUFA4L2 SNORD59B COQ10A GLI1 DDIT3 HSD17B6 PIP4K2C DCTN2 GLS2 PTGES3 R3HDM2 MIR1228 SNORA105C TIMELESS STAT2 DTX3 NABP2 SPRYD4 LRP1 INHBC BAZ2A OS9 RBMS2 ANKRD52 ZBTB39 MIR6758 GPR182 TAC3 ARHGAP9 |
nsv559035 | Chr.12:57467189 - 57476865 on Build GRCh38 | Loss | GLI1 ARHGAP9 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |