Assay Details
Target Gene Details
Entrez Gene ID: | 30 |
Gene Name: | acetyl-CoA acyltransferase 1 |
Gene Aliases: |
ACAA, PTHIO, THIO |
Location: |
Chr.3:38122710-38137242 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACAA1 | NM_001130410.1 | NP_001123882.1 | ||
NM_001607.3 | NP_001598.1 | |||
NR_024024.1 | ||||
XM_006713122.1 | XP_006713185.1 | |||
XM_006713123.2 | XP_006713186.1 | |||
XM_011533650.2 | XP_011531952.1 | |||
AF035295.1 | AAB88181.1 | |||
AK025138.1 | ||||
AK127051.1 | 5 | 1370 | ||
AK293716.1 | ||||
AK303251.1 | 8 | 1148 | ||
BC000635.2 | AAH00635.1 | |||
BC011977.1 | AAH11977.1 | |||
BC014474.1 | AAH14474.1 | |||
X12966.1 | CAA31412.1 | |||
X14813.1 | CAA32918.1 | |||
X51460.1 | CAA35825.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |