Assay Details
Target Gene Details
Entrez Gene ID: | 2050 |
Gene Name: | EPH receptor B4 |
Gene Aliases: |
HTK, MYK1, TYRO11 |
Location: |
Chr.7:100802565-100827521 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB4 | NM_004444.4 | 17 | 3766 | NP_004435.3 |
XM_017011816.1 | 17 | 3423 | XP_016867305.1 | |
AB209644.1 | 16 | 4476 | BAD92881.1 | |
AY056048.1 | 16 | 3204 | AAL14195.1 | |
BC004264.1 | 14 | 2851 | AAH04264.1 | |
BC052804.1 | 17 | 3443 | AAH52804.1 | |
U07695.1 | 17 | 3360 | AAA20598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470381 | Chr.7:100642568 - 100834358 on Build GRCh38 | Loss | EPO LOC105375429 POP7 TFR2 GNB2 ACTL6B ZAN EPHB4 GIGYF1 |
esv2759547 | Chr.7:100757867 - 101105965 on Build GRCh38 | Gain+Loss | TRIP6 ACHE MIR6875 MUC3A ZAN MUC12 EPHB4 SLC12A9 TRIM56 LOC102724094 MUC17 UFSP1 SRRT |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | EPO AP1S1 TRIP6 MIR6875 MUC12 PILRB NYAP1 TRIM56 GATS PCOLCE LOC102724094 MIR4653 PMS2P1 SERPINE1 MIR25 MIR93 AP4M1 CLDN15 STAG3L5P-PVRIG2P-PILRB MCM7 FBXO24 GNB2 MOGAT3 EPHB4 ZNHIT1 SLC12A9 PVRIG2P C7orf43 PLOD3 ZNF3 TAF6 TSC22D4 STAG3L5P UFSP1 NAT16 LRCH4 SRRT PPP1R35 COPS6 MIR106B FIS1 MUC3A AZGP1P1 GIGYF1 LOC105375429 GAL3ST4 STAG3 ZSCAN21 TFR2 SPDYE3 MOSPD3 ZKSCAN1 SAP25 PILRA ACHE AZGP1 ZASP POP7 ZCWPW1 MEPCE C7orf61 ACTL6B ZAN LAMTOR4 MBLAC1 PVRIG MIR4658 VGF MUC17 PCOLCE-AS1 MIR6840 CNPY4 GPC2 AGFG2 |
nsv607965 | Chr.7:100776465 - 100899511 on Build GRCh38 | Loss | TRIP6 ACHE MIR6875 UFSP1 ZAN EPHB4 SLC12A9 SRRT |
nsv1020837 | Chr.7:100715499 - 100911015 on Build GRCh38 | Loss | EPO TRIP6 ACHE MIR6875 UFSP1 ZAN EPHB4 SLC12A9 SRRT |
nsv951388 | Chr.7:100741378 - 100838178 on Build GRCh38 | Deletion | ZAN EPHB4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |