Assay Details
Target Gene Details
Entrez Gene ID: | 124989 |
Gene Name: | EF-hand calcium binding domain 13 |
Gene Aliases: |
C17orf57 |
Location: |
Chr.17:47323961-47441312 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EFCAB13 | NM_001195192.1 | NP_001182121.1 | ||
NM_152347.4 | NP_689560.3 | |||
BC036407.2 | AAH36407.1 | |||
BC037876.1 | AAH37876.1 | |||
BX471329.1 |