Assay Details
Target Gene Details
Entrez Gene ID: | 8711 |
Gene Name: | tyrosine kinase non receptor 1 |
Gene Aliases: |
- |
Location: |
Chr.17:7380559-7389774 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNK1 | NM_001251902.1 | NP_001238831.1 | ||
NM_003985.4 | NP_003976.2 | |||
XM_011524045.2 | XP_011522347.1 | |||
XM_011524046.2 | XP_011522348.1 | |||
XM_011524047.2 | XP_011522349.1 | |||
XM_011524048.1 | XP_011522350.1 | |||
AK225403.1 | ||||
AK310684.1 | ||||
AK311063.1 | 2 | 567 | ||
BC035782.1 | AAH35782.1 | |||
DA089305.1 | ||||
DB287365.1 | ||||
U43408.1 | AAC50427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion | ALOX12-AS1 TMEM95 ASGR2 MIR195 TNK1 ATP1B2 SNORA48 TMEM256 CHRNB1 C17orf49 EIF4A1 TEKT1 POLR2A TMEM256-PLSCR3 TMEM102 MIR497 YBX2 ALOX12 SOX15 CLDN7 SLC2A4 CTDNEP1 LOC107983988 SLC16A13 RNASEK-C17orf49 DVL2 GABARAP SLC16A11 ZBTB4 SENP3 SLC35G6 PLSCR3 MIR324 ACADVL FXR2 ACAP1 GPS2 ELP5 SHBG KCTD11 BCL6B FGF11 FBXO39 TNFSF13 DLG4 SAT2 RNASEK SPEM1 TNFSF12 NLGN2 ASGR1 LOC100996842 CD68 CLEC10A SNORD10 C17orf74 MIR497HG SNORA67 NEURL4 TNFSF12-TNFSF13 MPDU1 PHF23 ALOX12P2 SENP3-EIF4A1 EIF5A |
nsv523672 | Chr.17:7379825 - 7549660 on Build GRCh38 | Loss | SPEM1 ZBTB4 SLC35G6 TNFSF12 NLGN2 PLSCR3 TNK1 POLR2A TMEM256-PLSCR3 TMEM102 C17orf74 TNFSF12-TNFSF13 LOC107983988 FGF11 TMEM256 CHRNB1 |
nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion | DVL2 TMEM95 GABARAP PLSCR3 MIR324 TNK1 ACADVL ACAP1 GPS2 ELP5 KCTD11 FGF11 TMEM256 CHRNB1 DLG4 SPEM1 NLGN2 ASGR1 TMEM256-PLSCR3 TMEM102 YBX2 CLDN7 C17orf74 SLC2A4 CTDNEP1 NEURL4 LOC107983988 PHF23 EIF5A |
nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss | ALOX12-AS1 TMEM95 ASGR2 MIR195 TNK1 ATP1B2 SNORA48 TMEM256 CHRNB1 C17orf49 EIF4A1 POLR2A TMEM256-PLSCR3 TMEM102 MIR497 YBX2 ALOX12 SOX15 CLDN7 SLC2A4 CTDNEP1 LOC107983988 SLC16A13 RNASEK-C17orf49 WRAP53 DVL2 GABARAP SLC16A11 ZBTB4 SENP3 SLC35G6 PLSCR3 MIR324 ACADVL FXR2 ACAP1 GPS2 ELP5 SHBG KCTD11 BCL6B FGF11 TNFSF13 DLG4 SAT2 RNASEK SPEM1 TNFSF12 NLGN2 ASGR1 TP53 LOC100996842 CD68 CLEC10A SNORD10 C17orf74 MIR497HG SNORA67 NEURL4 TNFSF12-TNFSF13 MPDU1 PHF23 SENP3-EIF4A1 EIF5A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |