Assay Details
Target Gene Details
Entrez Gene ID: | 2359 |
Gene Name: | formyl peptide receptor 3 |
Gene Aliases: |
FML2_HUMAN, FMLP-R-II, FMLPY, FPRH1, FPRH2, FPRL2, RMLP-R-I |
Location: |
Chr.19:51795146-51826207 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FPR3 | NM_002030.4 | 2 | 656 | NP_002021.3 |
XM_011526687.2 | 2 | 547 | XP_011524989.1 | |
AK123030.1 | 3 | 599 | ||
AK292541.1 | 3 | 645 | ||
BC059388.1 | 2 | 639 | AAH59388.1 | |
BC069070.1 | 1 | 460 | AAH69070.1 | |
BC069593.1 | 1 | 460 | AAH69593.1 | |
BC069812.1 | 1 | 460 | AAH69812.1 | |
CB996360.1 | 2 | 666 | ||
M76673.1 | 1 | 460 | AAA58482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv425n27 | Chr.19:51784199 - 52104512 on Build GRCh38 | Gain | ZNF649 ZNF649-AS1 LOC100419706 FPR3 ZNF841 ZNF350-AS1 ZNF350 ZNF432 ZNF615 ZNF614 ZNF577 ZNF613 |
dgv1067e212 | Chr.19:51768056 - 52117340 on Build GRCh38 | Gain | ZNF350-AS1 ZNF350 ZNF615 ZNF616 ZNF577 ZNF649 FPR2 ZNF649-AS1 LOC100419706 FPR3 ZNF841 ZNF432 ZNF614 ZNF613 |
esv3644680 | Chr.19:51785765 - 51953257 on Build GRCh38 | Gain | ZNF649 ZNF649-AS1 LOC100419706 FPR3 ZNF350-AS1 ZNF577 ZNF613 |
dgv143e55 | Chr.19:51724663 - 52127682 on Build GRCh38 | Gain | ZNF350-AS1 ZNF350 ZNF615 FPR1 ZNF616 ZNF577 ZNF649 FPR2 ZNF649-AS1 LOC100419706 FPR3 ZNF841 ZNF432 ZNF614 ZNF613 |
dgv3625n100 | Chr.19:51722260 - 51847643 on Build GRCh38 | Gain | HAS1 FPR2 FPR3 FPR1 |
esv2758767 | Chr.19:51488642 - 52101372 on Build GRCh38 | Gain+Loss | ZNF350-AS1 ZNF350 SPACA6P-AS LOC339352 FPR1 ZNF577 MIRLET7E HAS1 SIGLEC14 FPR2 FPR3 SIGLEC6 ZNF615 CEACAM18 MIR99B ZNF649 ZNF649-AS1 SIGLEC12 LINC01530 LOC100419706 SPACA6 ZNF841 ZNF432 SIGLEC5 MIR125A ZNF614 ZNF613 ZNF175 |
nsv580001 | Chr.19:51798407 - 52013985 on Build GRCh38 | Gain | ZNF649 ZNF649-AS1 LOC100419706 FPR3 ZNF350-AS1 ZNF350 ZNF615 ZNF614 ZNF577 ZNF613 |
dgv3626n100 | Chr.19:51746647 - 52125358 on Build GRCh38 | Gain | ZNF350-AS1 ZNF350 ZNF615 FPR1 ZNF616 ZNF577 ZNF649 FPR2 ZNF649-AS1 LOC100419706 FPR3 ZNF841 ZNF432 ZNF614 ZNF613 |
dgv6523n54 | Chr.19:51767689 - 52118391 on Build GRCh38 | Gain | ZNF350-AS1 ZNF350 ZNF615 ZNF616 ZNF577 ZNF649 FPR2 ZNF649-AS1 LOC100419706 FPR3 ZNF841 ZNF432 ZNF614 ZNF613 |
nsv458733 | Chr.19:51722514 - 51856730 on Build GRCh38 | Gain | HAS1 FPR2 FPR3 FPR1 ZNF577 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |