Assay Details
Target Gene Details
Entrez Gene ID: | 83986 |
Gene Name: | family with sequence similarity 234 member A |
Gene Aliases: |
C16orf9, ITFG3, gs19 |
Location: |
Chr.16:234546-269943 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM234A | NR_104317.1 | 13 | 1976 | |
AK057165.1 | 14 | 1818 | BAB71374.1 | |
AK294581.1 | 13 | 1688 |
Target Gene Details
Entrez Gene ID: | 8786 |
Gene Name: | regulator of G-protein signaling 11 |
Gene Aliases: |
RS11 |
Location: |
Chr.16:268301-275943 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS11 | NM_001286485.1 | 16 | 1819 | NP_001273414.1 |
NM_001286486.1 | 15 | 1687 | NP_001273415.1 | |
NM_003834.2 | 17 | 1863 | NP_003825.1 | |
NM_183337.2 | 17 | 1852 | NP_899180.1 | |
XM_011522719.2 | 18 | 1946 | XP_011521021.1 | |
XM_011522720.2 | 18 | 1886 | XP_011521022.1 | |
AB209582.1 | 9 | 3760 | BAD92819.1 | |
AF035153.1 | 17 | 1823 | AAC69175.1 | |
AF035154.1 | 17 | 1834 | AAC69176.1 | |
AK091701.1 | 15 | 1684 | ||
BC141803.1 | 17 | 1827 | ||
CK299324.1 | 1 | 571 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | LOC105371049 STUB1 MIR662 DDX11L10 PIGQ FAM234A NME3 HBA1 CCDC78 PRR25 CAPN15 C1QTNF8 LMF1-AS1 SSTR5 RNF151 WDR90 TPSAB1 POLR3K RHBDL1 BAIAP3 MSLN DECR2 PDIA2 ABCA17P NPRL3 NTN3 SNORA64 TPSG1 ABCA3 SNORA78 SPSB3 BRICD5 MIR3176 WDR24 WASIR2 CHTF18 SOX8 HBM HAGH FBXL16 CCNF SLC9A3R2 RHOT2 TBC1D24 MIR3180-5 MRPL28 IFT140 MIR1225 CEMP1 RAB11FIP3 ARHGDIG TSC2 RAB26 ATP6V0C MEIOB RPL23AP5 TMEM8A ERVK13-1 LINC00254 UNKL C16orf59 LOC100287175 LOC105371046 CCDC154 FAHD1 LOC106660606 HN1L RNPS1 MIR6859-4 MPG E4F1 C16orf91 IGFALS WFIKKN1 KCTD5 MIR6511B1 NUBP2 LOC100134368 PRSS27 HBQ1 MIR940 MLST8 MRPS34 EME2 ZNF598 TPSB2 UBE2I RPUSD1 GNPTG RHBDF1 CLCN7 TSR3 NDUFB10 MIR3177 PRSS29P NPW LUC7L LOC105371047 SNHG19 ECI1 PRR35 GNG13 PDPK1 LOC105371038 HBZ MIR6768 NOXO1 FLJ42627 HS3ST6 SYNGR3 METTL26 PTX4 METRN CACNA1H NME4 MIR3677 LOC729652 SNRNP25 LOC105371184 TMEM204 GFER PGP CASKIN1 SNORA10 NARFL SSTR5-AS1 LOC107987233 MAPK8IP3 SNORD60 LMF1 FAM173A RPL3L LOC101929280 AXIN1 MIR5587 HAGHL TELO2 MIR4717 TRAF7 JMJD8 PKD1 RGS11 SNHG9 LOC105371045 LINC00235 LOC652276 LOC107984876 DNASE1L2 MCRIP2 TBL3 NTHL1 AMDHD2 MIR4516 RPS2 MIR6767 TPSD1 NHLRC4 MSRB1 MIR3178 RAB40C CRAMP1 HBA2 |
esv3570031 | Chr.16:243389 - 278107 on Build GRCh38 | Gain | RGS11 FAM234A |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | RAB11FIP3 AXIN1 LOC100134368 ARHGDIG RGS11 RPL23AP5 TMEM8A DECR2 NME4 PDIA2 FAM234A MRPL28 |
nsv827477 | Chr.16:262247 - 297639 on Build GRCh38 | Gain | AXIN1 ARHGDIG RGS11 PDIA2 FAM234A |
esv3637561 | Chr.16:261769 - 271795 on Build GRCh38 | Loss | RGS11 FAM234A |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | RAB11FIP3 AXIN1 LOC100134368 ARHGDIG RGS11 RPL23AP5 TMEM8A DECR2 NME4 PDIA2 FAM234A MRPL28 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |