Assay Details
Target Gene Details
Entrez Gene ID: | 4240 |
Gene Name: | milk fat globule-EGF factor 8 protein |
Gene Aliases: |
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47 |
Location: |
Chr.15:88898683-88913469 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFGE8 | NM_001114614.2 | 6 | 968 | NP_001108086.1 |
NM_001310319.1 | 5 | 836 | NP_001297248.1 | |
NM_001310320.1 | 7 | 1021 | NP_001297249.1 | |
NM_001310321.1 | 6 | 1080 | NP_001297250.1 | |
NM_005928.3 | 6 | 968 | NP_005919.2 | |
AK095908.1 | 7 | 931 | ||
AK222735.1 | 6 | 878 | BAD96455.1 | |
AK223182.1 | 6 | 878 | BAD96902.1 | |
AK304627.1 | 5 | 746 | ||
AK312640.1 | 6 | 878 | ||
AY141974.1 | 6 | 887 | AAN08508.1 | |
BC003610.2 | 6 | 861 | AAH03610.1 | |
BT006948.1 | 6 | 818 | AAP35594.1 | |
BX537974.1 | 6 | 842 | CAD97938.1 | |
S56151.1 | 3 | 311 | AAB19771.1 | |
U58516.1 | 6 | 878 | AAC50549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1653 | Chr.15:88870409 - 88915341 on Build GRCh38 | Deletion | ACAN HAPLN3 MFGE8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |