Assay Details
Target Gene Details
Entrez Gene ID: | 51272 |
Gene Name: | Bet1 golgi vesicular membrane trafficking protein like |
Gene Aliases: |
BET1L1, GOLIM3, GS15, HSPC197 |
Location: |
Chr.11:202924-207422 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BET1L | NM_001098787.1 | NP_001092257.1 | ||
NM_016526.4 | NP_057610.2 | |||
AF234160.1 | AAF37877.1 | |||
AK022958.1 | ||||
BC008971.1 | AAH08971.1 | |||
BC032779.2 | AAH32779.1 | |||
DA875317.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3625061 | Chr.11:195766 - 233956 on Build GRCh38 | Gain | ODF3 RIC8A BET1L MIR6743 SIRT3 |
esv3625060 | Chr.11:194412 - 252166 on Build GRCh38 | Gain | ODF3 RIC8A BET1L MIR6743 SCGB1C1 PSMD13 SIRT3 |
nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain | ODF3 PGGHG RIC8A BET1L IFITM3 MIR6743 IFITM5 PSMD13 IFITM2 IFITM1 NLRP6 SIRT3 |
dgv1011n100 | Chr.11:198510 - 275185 on Build GRCh38 | Gain | ODF3 RIC8A BET1L MIR6743 PSMD13 SIRT3 |
esv22231 | Chr.11:60059 - 216243 on Build GRCh38 | Gain | LINC01001 ODF3 RIC8A BET1L MIR6743 SCGB1C1 LOC388572 SIRT3 |
nsv951267 | Chr.11:196001 - 206600 on Build GRCh38 | Deletion | ODF3 BET1L |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |