Assay Details
Target Gene Details
Entrez Gene ID: | 80208 |
Gene Name: | spastic paraplegia 11 (autosomal recessive) |
Gene Aliases: |
ALS5, CMT2X, KIAA1840 |
Location: |
Chr.15:44562696-44663678 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPG11 | NM_001160227.1 | 29 | 5104 | NP_001153699.1 |
NM_025137.3 | 29 | 5104 | NP_079413.3 | |
XM_006720700.1 | 29 | 5104 | XP_006720763.1 | |
XM_017022634.1 | 29 | 5104 | XP_016878123.1 | |
XM_017022636.1 | 18 | 2913 | XP_016878125.1 | |
AB058743.2 | 29 | 5074 | BAB47469.2 | |
AB470308.1 | 29 | 5073 | ||
AB470309.1 | 29 | 5073 | ||
AB470310.1 | 29 | 5073 | ||
AK091176.1 | 9 | 1463 | BAC03600.1 | |
BC150640.1 | 29 | 5101 | ||
BC153879.1 | 29 | 5074 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |