Assay Details
Target Gene Details
Entrez Gene ID: | 3981 |
Gene Name: | DNA ligase 4 |
Gene Aliases: |
LIG4S |
Location: |
Chr.13:108207442-108218368 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIG4 | NM_001098268.1 | 2 | 1571 | NP_001091738.1 |
NM_002312.3 | 2 | 1609 | NP_002303.2 | |
NM_206937.1 | 3 | 1488 | NP_996820.1 | |
XM_005254056.1 | 3 | 1453 | XP_005254113.1 | |
XM_005254057.4 | 3 | 1623 | XP_005254114.1 | |
XM_005254058.3 | 2 | 3069 | XP_005254115.1 | |
XM_006719951.3 | 3 | 3328 | XP_006720014.1 | |
XM_006719952.1 | 4 | 1921 | XP_006720015.1 | |
XM_011521091.2 | 4 | 1950 | XP_011519393.1 | |
XM_011521092.2 | 3 | 1526 | XP_011519394.1 | |
XM_017020563.1 | 3 | 3170 | XP_016876052.1 | |
XM_017020564.1 | 2 | 1596 | XP_016876053.1 | |
XM_017020565.1 | 4 | 1967 | XP_016876054.1 | |
XM_017020566.1 | 4 | 1870 | XP_016876055.1 | |
XM_017020567.1 | 3 | 1524 | XP_016876056.1 | |
XM_017020568.1 | 3 | 3146 | XP_016876057.1 | |
XM_017020569.1 | 4 | 3591 | XP_016876058.1 | |
XM_017020570.1 | 4 | 1870 | XP_016876059.1 | |
XM_017020571.1 | 4 | 3675 | XP_016876060.1 | |
XM_017020572.1 | 3 | 3062 | XP_016876061.1 | |
XM_017020573.1 | 2 | 1372 | XP_016876062.1 | |
AK292419.1 | 3 | 1397 | ||
AK314844.1 | 3 | 1453 | ||
BC037491.2 | 3 | 1488 | AAH37491.1 | |
BX648833.1 | 2 | 1572 | ||
X83441.1 | 2 | 1609 | CAA58467.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |