Assay Details
Target Gene Details
Entrez Gene ID: | 51126 |
Gene Name: | N(alpha)-acetyltransferase 20, NatB catalytic subunit |
Gene Aliases: |
NAT3, NAT3P, NAT5, NAT5P, dJ1002M8.1 |
Location: |
Chr.20:20017290-20033629 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA20 | NM_016100.4 | NP_057184.1 | ||
NM_181527.3 | 1 | 157 | NP_852668.1 | |
NM_181528.3 | NP_852669.1 | |||
AF085355.1 | AAD40190.1 | |||
AK311819.1 | ||||
AL136641.1 | CAB66576.1 | |||
BC005181.1 | AAH05181.1 | |||
BC008446.1 | AAH08446.1 | |||
BG548527.1 | ||||
BX338359.1 | 6 | 955 | ||
CA867744.1 | ||||
CB959658.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |