Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYOU1 | NM_001130991.2 | 26 | 4243 | NP_001124463.1 |
NM_006389.4 | 26 | 4376 | NP_006380.1 | |
XM_005271392.3 | 26 | 4240 | XP_005271449.1 | |
XM_005271393.3 | 26 | 4220 | XP_005271450.1 | |
XM_005271394.3 | 26 | 4287 | XP_005271451.1 | |
XM_017017095.1 | 26 | 4275 | XP_016872584.1 | |
XM_017017096.1 | 26 | 4293 | XP_016872585.1 | |
XM_017017097.1 | 26 | 4216 | XP_016872586.1 | |
AA580314.1 | 1 | 287 | ||
AK127324.1 | 25 | 4248 | ||
BC004560.2 | 7 | 1309 | AAH04560.1 | |
BC072436.1 | 19 | 2800 | AAH72436.1 | |
CR749610.1 | 4 | 1313 | CAH18405.1 | |
U65785.1 | 26 | 4244 | AAC50947.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952745 | Chr.11:119041291 - 119047288 on Build GRCh38 | Deletion | HYOU1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |