Assay Details
Target Gene Details
Entrez Gene ID: | 2322 |
Gene Name: | fms related tyrosine kinase 3 |
Gene Aliases: |
CD135, FLK-2, FLK2, STK1 |
Location: |
Chr.13:28003274-28100592 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLT3 | NM_004119.2 | NP_004110.2 | ||
NR_130706.1 | ||||
XM_011535015.2 | XP_011533317.1 | |||
XM_011535017.2 | XP_011533319.1 | |||
XM_011535018.2 | XP_011533320.1 | |||
XM_017020486.1 | XP_016875975.1 | |||
XM_017020487.1 | XP_016875976.1 | |||
XM_017020488.1 | XP_016875977.1 | |||
XM_017020489.1 | XP_016875978.1 | |||
BC126350.1 | ||||
BC144039.1 | ||||
BC144040.1 | ||||
L36162.1 | AAA35487.1 | |||
U02687.1 | AAA18947.1 | |||
Z26652.1 | CAA81393.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |