Assay Details
Target Gene Details
Entrez Gene ID: | 1271 |
Gene Name: | ciliary neurotrophic factor receptor |
Gene Aliases: |
- |
Location: |
Chr.9:34551432-34590386 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTFR | NM_001207011.1 | 4 | 295 | NP_001193940.1 |
NM_001842.4 | 3 | 308 | NP_001833.1 | |
NM_147164.2 | 4 | 419 | NP_671693.1 | |
XM_017014260.1 | 5 | 635 | XP_016869749.1 | |
XM_017014261.1 | 5 | 608 | XP_016869750.1 | |
XM_017014262.1 | 5 | 552 | XP_016869751.1 | |
XM_017014263.1 | 4 | 393 | XP_016869752.1 | |
XM_017014264.1 | 4 | 937 | XP_016869753.1 | |
XM_017014265.1 | 3 | 253 | XP_016869754.1 | |
AL537375.3 | 4 | 294 | ||
BC001492.2 | 3 | 265 | AAH01492.1 | |
BI545014.1 | 4 | 418 | ||
BI829871.1 | 3 | 309 | ||
BT019824.1 | 2 | 125 | AAV38627.1 | |
HQ628632.1 | 2 | 125 | ||
M73238.1 | 4 | 388 | AAA35707.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain | CNTFR DCAF12 LOC730098 LOC114224 SIGMAR1 IL11RA SNORD121B ENHO FAM205A UBAP2 DNAI1 SNORD121A RPP25L NUDT2 CNTFR-AS1 UBE2R2 FAM219A KIF24 CCL19 DCTN3 GALT CCL27 UBAP1 KIAA1161 ARID3C C9orf24 CCL21 |
nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion | DCTN3 SIGMAR1 RPP25L IL11RA CNTFR GALT CCL27 CNTFR-AS1 LOC730098 ARID3C |
dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss | DCTN3 SIGMAR1 RPP25L IL11RA CNTFR GALT CCL27 CNTFR-AS1 ARID3C |
nsv831543 | Chr.9:34431291 - 34615384 on Build GRCh38 | Loss | DCTN3 DNAI1 RPP25L CNTFR ENHO CNTFR-AS1 FAM219A |
nsv951699 | Chr.9:34553903 - 34574802 on Build GRCh38 | Deletion | CNTFR CNTFR-AS1 |
nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain | DCTN3 SIGMAR1 RPP25L IL11RA CNTFR GALT CCL27 CNTFR-AS1 LOC730098 ARID3C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |