Assay Details
Target Gene Details
Entrez Gene ID: | 2997 |
Gene Name: | glycogen synthase 1 |
Gene Aliases: |
GSY, GYS |
Location: |
Chr.19:48968125-48993353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GYS1 | NM_001161587.1 | NP_001155059.1 | ||
NM_002103.4 | NP_002094.2 | |||
NR_027763.1 | ||||
AK223579.1 | BAD97299.1 | |||
AK291672.1 | ||||
AK294349.1 | ||||
AK300369.1 | ||||
AK302738.1 | ||||
AK303712.1 | ||||
BC002617.2 | AAH02617.1 | |||
BC003182.1 | AAH03182.1 | |||
BC007688.2 | AAH07688.1 | |||
J04501.1 | AAA88046.1 | |||
U32573.1 | AAB60385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | FTL KCNA7 ADM5 CA11 TBC1D17 SNAR-D MAMSTR PNKP SNAR-A11 SNAR-A9 SEC1P SNORD33 NTN5 CPT1C RRAS MIR5088 FUT2 SNORD34 HRC DBP SCAF1 SNAR-C4 CYTH2 BAX CARD8 AP2A1 LMTK3 PTOV1-AS2 SNAR-A1 FCGRT SNORD35B CD37 SLC6A16 LHB GFY SNRNP70 ZNF114 SNAR-B2 IL4I1 MIR6798 SNORD35A NUCB1-AS1 SNAR-A13 SIGLEC16 NOSIP RPL13A TSKS ZNF473 PTOV1-AS1 SPHK2 TEAD2 KDELR1 LOC101059948 LIN7B CCDC155 MIR4750 SNAR-G1 TMEM143 C19orf73 LIG1 CCDC114 VRK3 SNAR-B1 LOC105372430 MIR150 PTOV1 SLC17A7 MIR4324 SNAR-A3 CGB8 IRF3 LOC105447645 SNORD32A MED25 DKKL1 NUCB1 PRR12 CABP5 SNAR-A12 FLJ26850 CARD8-AS1 PLEKHA4 MIR4751 SULT2B1 SNAR-A4 SNAR-G2 NUP62 RPL18 SNAR-C5 DHDH CGB3 MIR6800 SNAR-C1 GRIN2D PIH1D1 TULP2 GYS1 SPACA4 BCL2L12 MIR4749 SNAR-A8 AKT1S1 FGF21 RUVBL2 SNAR-C2 RCN3 CGB5 PPFIA3 SNAR-A7 CGB2 SNAR-A10 FAM83E SIGLEC11 SYNGR4 PRMT1 FUZ ALDH16A1 PTH2 RASIP1 PRRG2 EMP3 MIR6799 GRWD1 BCAT2 SNAR-A5 IZUMO1 LOC101928295 SNAR-A6 NTF4 BSPH1 SNAR-C3 ATF5 FUT1 LOC100287477 RPS11 SNAR-A14 SNAR-A2 FLT3LG CGB7 ELSPBP1 PLA2G4C TRPM4 CGB1 HSD17B14 PPP1R15A PLA2G4C-AS1 C19orf68 KCNJ14 |
dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain | FTL MIR6798 GYS1 LHB BAX LOC101059948 RUVBL2 |
nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss | CGB8 NTF4 FTL MIR6798 KCNA7 CGB5 PPFIA3 SNAR-G1 DHDH CGB3 BAX NUCB1-AS1 CGB2 C19orf73 NUCB1 CGB7 GYS1 LHB LOC101059948 CGB1 SNRNP70 LIN7B RUVBL2 SNAR-G2 |
nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion | CGB8 NTF4 FTL MIR6798 KCNA7 CGB5 SNAR-G1 CGB3 BAX CGB2 CGB7 GYS1 LHB LOC101059948 CGB1 SNRNP70 RUVBL2 SNAR-G2 |
nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss | FTL TULP2 GYS1 DHDH BAX NUCB1-AS1 PLEKHA4 PPP1R15A NUCB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |