Assay Details
Target Gene Details
Entrez Gene ID: | 83706 |
Gene Name: | fermitin family member 3 |
Gene Aliases: |
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF |
Location: |
Chr.11:64206665-64223891 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FERMT3 | NM_031471.5 | NP_113659.3 | ||
NM_178443.2 | NP_848537.1 | |||
XM_011545294.2 | XP_011543596.1 | |||
XM_011545295.2 | XP_011543597.1 | |||
XM_017018398.1 | XP_016873887.1 | |||
XM_017018399.1 | XP_016873888.1 | |||
AK093719.1 | BAC04220.1 | |||
AY093951.1 | AAM19736.1 | |||
AY093952.1 | AAM19737.1 | |||
BC004347.1 | AAH04347.2 | |||
BC013366.2 | AAH13366.1 | |||
BC015584.2 | AAH15584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | DNAJC4 GPR137 VEGFB FERMT3 NUDT22 PPP1R14B PLCB3 FKBP2 BAD TRPT1 STIP1 LOC105369340 MACROD1 |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | PRDX5 VEGFB KCNK4 FERMT3 CCDC88B NUDT22 MIR7155 BAD TRPT1 STIP1 KCNK4-TEX40 DNAJC4 GPR137 FLRT1 TEX40 OTUB1 NAA40 ESRRA PPP1R14B PLCB3 FKBP2 TRMT112 COX8A LOC105369340 MACROD1 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | DNAJC4 GPR137 PRDX5 VEGFB KCNK4 TEX40 FERMT3 NUDT22 ESRRA PPP1R14B PLCB3 FKBP2 TRMT112 BAD TRPT1 STIP1 KCNK4-TEX40 LOC105369340 |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | DNAJC4 PPP1R14B PLCB3 FKBP2 VEGFB TRPT1 STIP1 LOC105369340 FERMT3 NUDT22 |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | DNAJC4 GPR137 PRDX5 VEGFB KCNK4 TEX40 FERMT3 CCDC88B NUDT22 MIR7155 MIR1237 ESRRA PPP1R14B PLCB3 FKBP2 TRMT112 BAD TRPT1 KCNK4-TEX40 LOC105369340 RPS6KA4 |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | DNAJC4 GPR137 VEGFB FERMT3 NUDT22 PPP1R14B PLCB3 FKBP2 BAD TRPT1 STIP1 LOC105369340 MACROD1 |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | DNAJC4 GPR137 PRDX5 FLRT1 VEGFB KCNK4 TEX40 FERMT3 NUDT22 ESRRA PPP1R14B PLCB3 FKBP2 TRMT112 BAD TRPT1 STIP1 KCNK4-TEX40 LOC105369340 MACROD1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |