Assay Details
Target Gene Details
Entrez Gene ID: | 7009 |
Gene Name: | transmembrane BAX inhibitor motif containing 6 |
Gene Aliases: |
BAXI1, BI-1, TEGT |
Location: |
Chr.12:49741510-49764934 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMBIM6 | NM_001098576.1 | 4 | 723 | NP_001092046.1 |
NM_003217.2 | 4 | 353 | NP_003208.2 | |
XM_005269126.3 | 5 | 625 | XP_005269183.1 | |
AF033095.1 | 4 | 283 | AAB87479.1 | |
AK124849.1 | 4 | 1048 | ||
AK130050.1 | 4 | 306 | ||
AK130462.1 | 2 | 175 | ||
AK225804.1 | 4 | 306 | ||
AK299587.1 | 4 | 307 | ||
AK304577.1 | 4 | 301 | ||
AK312238.1 | 4 | 289 | ||
AY736129.1 | 3 | 221 | AAU29521.1 | |
BC000916.1 | 4 | 286 | AAH00916.1 | |
BC036203.1 | 4 | 361 | ||
DA476870.1 | 4 | 354 | ||
DC319835.1 | 4 | 483 | ||
DC349385.1 | ||||
X75861.1 | 3 | 261 | CAA53472.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |