Assay Details
Target Gene Details
Entrez Gene ID: | 22933 |
Gene Name: | sirtuin 2 |
Gene Aliases: |
SIR2, SIR2L, SIR2L2 |
Location: |
Chr.19:38878555-38899862 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRT2 | NM_001193286.1 | 13 | 1514 | NP_001180215.1 |
NM_012237.3 | 16 | 1674 | NP_036369.2 | |
NM_030593.2 | 15 | 1666 | NP_085096.1 | |
NR_034146.1 | 14 | 1597 | ||
XM_006723111.1 | 17 | 1622 | XP_006723174.1 | |
XM_011526654.1 | 15 | 1559 | XP_011524956.1 | |
XM_011526655.1 | 13 | 1374 | XP_011524957.1 | |
XM_017026500.1 | 15 | 1598 | XP_016881989.1 | |
AB009274.1 | 15 | 1387 | ||
AF083107.2 | 16 | 1572 | AAD40850.2 | |
AF095714.1 | 15 | 1445 | AAD45971.1 | |
AF131800.1 | 12 | 1169 | AAD20046.1 | |
AF160214.1 | 15 | 1426 | AAF67015.1 | |
AJ505014.1 | 13 | 2143 | CAD43717.1 | |
AK025876.1 | 15 | 1452 | ||
AK054642.1 | 16 | 1505 | ||
AK131390.1 | 5 | 4338 | ||
AK225824.1 | 15 | 1448 | ||
AY030277.1 | 15 | 1427 | AAK51133.1 | |
BC003012.1 | 15 | 1517 | AAH03012.1 | |
BC003547.1 | 15 | 1517 | AAH03547.1 | |
BF346568.1 | 1 | 254 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160616 | Chr.19:38839331 - 38903590 on Build GRCh38 | Duplication | HNRNPL RINL NFKBIB SIRT2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |