Assay Details
Target Gene Details
Entrez Gene ID: | 2109 |
Gene Name: | electron transfer flavoprotein beta subunit |
Gene Aliases: |
FP585, MADD |
Location: |
Chr.19:51345155-51366418 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ETFB | NM_001014763.1 | 5 | 1238 | NP_001014763.1 |
NM_001985.2 | 6 | 853 | NP_001976.1 | |
AF370381.1 | 5 | 1220 | AAQ15217.1 | |
AK055285.1 | ||||
AK291881.1 | ||||
AL833205.1 | 5 | 3514 | ||
BC093961.1 | AAH93961.1 | |||
BC093963.1 | AAH93963.1 | |||
BG281641.1 | 5 | 671 | ||
CR456827.1 | CAG33108.1 | |||
X71129.1 | 6 | 788 | CAA50441.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |