Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | 8 | 1546 | NP_064536.2 |
XM_011511499.1 | 8 | 1163 | XP_011509801.1 | |
AF270647.1 | 8 | 1473 | AAF81750.1 | |
AK023234.1 | 7 | 805 | ||
AK054615.1 | 8 | 912 | ||
AK094925.1 | 2 | 1118 | ||
AK096666.1 | ||||
AK291190.1 | 8 | 928 | ||
BC007894.2 | 9 | 1304 | AAH07894.1 | |
BC008894.2 | 8 | 1529 | AAH08894.1 | |
BC009998.2 | 9 | 1304 | AAH09998.1 | |
BC012774.2 | 8 | 1496 | AAH12774.1 | |
BC012793.2 | 8 | 1510 | AAH12793.1 | |
BC013279.2 | 8 | 1529 | AAH13279.1 | |
BC014076.2 | 8 | 1506 | AAH14076.1 | |
BC014274.2 | 8 | 1525 | AAH14274.3 | |
BC032106.1 | 8 | 1520 | AAH32106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)