Assay Details
Target Gene Details
Entrez Gene ID: | 688 |
Gene Name: | Kruppel like factor 5 |
Gene Aliases: |
BTEB2, CKLF, IKLF |
Location: |
Chr.13:73054976-73077542 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF5 | NM_001286818.1 | 2 | 722 | NP_001273747.1 |
NM_001730.4 | 2 | 1336 | NP_001721.2 | |
AB030824.1 | 2 | 1002 | BAA96461.1 | |
AF132818.1 | 2 | 1336 | AAF18307.1 | |
AF287272.1 | 2 | 1111 | AAF88068.1 | |
AK296900.1 | 3 | 1066 | ||
AK302280.1 | 2 | 722 | ||
AK304284.1 | 3 | 1030 | ||
BC007695.1 | 2 | 1114 | AAH07695.1 | |
BC042131.1 | 2 | 973 | AAH42131.1 | |
BT006641.1 | 2 | 800 | AAP35287.1 | |
D14520.1 | 2 | 645 | BAA03393.1 | |
EF208215.1 | 2 | 722 | ||
HF546203.1 | 2 | 601 | ||
HF546204.1 | ||||
HQ628639.1 | 3 | 609 | ||
HQ628640.1 | 3 | 753 | ||
HQ628641.1 | 3 | 779 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |