Assay Details
Target Gene Details
Entrez Gene ID: | 79948 |
Gene Name: | phospholipid phosphatase related 3 |
Gene Aliases: |
LPPR3, LPR3, PRG-2, PRG2 |
Location: |
Chr.19:812488-824244 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPPR3 | NM_001270366.1 | 2 | 89 | NP_001257295.1 |
NM_024888.2 | 2 | 89 | NP_079164.1 | |
XM_011528317.2 | 2 | 681 | XP_011526619.1 | |
AF357888.1 | 1 | 433 | AAO85401.1 | |
AF541282.1 | 2 | 89 | AAP57771.1 | |
AL136596.1 | 2 | 89 | CAB66531.1 | |
AY304517.1 | 2 | 89 | AAP72154.1 | |
AY436785.1 | 2 | 88 | AAR10818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828357 | Chr.19:817381 - 823632 on Build GRCh38 | Loss |
![]() |
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv578172 | Chr.19:811414 - 841398 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv578167 | Chr.19:797079 - 850574 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)