Assay Details
Target Gene Details
Entrez Gene ID: | 10525 |
Gene Name: | hypoxia up-regulated 1 |
Gene Aliases: |
GRP-170, Grp170, HSP12A, ORP-150, ORP150 |
Location: |
Chr.11:119044185-119057246 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HYOU1 | NM_001130991.2 | NP_001124463.1 | ||
NM_006389.4 | NP_006380.1 | |||
XM_005271392.3 | XP_005271449.1 | |||
XM_005271393.3 | XP_005271450.1 | |||
XM_005271394.3 | XP_005271451.1 | |||
XM_017017095.1 | XP_016872584.1 | |||
XM_017017096.1 | XP_016872585.1 | |||
XM_017017097.1 | XP_016872586.1 | |||
AK127324.1 | ||||
AK222756.1 | BAD96476.1 | |||
AK294905.1 | ||||
AK297260.1 | ||||
AK299648.1 | ||||
AK301514.1 | ||||
AK304264.1 | ||||
AK314178.1 | ||||
BC004560.2 | AAH04560.1 | |||
BC072436.1 | AAH72436.1 | |||
DQ350134.1 | ABC75106.1 | |||
DQ372932.1 | ABD14370.1 | |||
U65785.1 | AAC50947.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |