Assay Details
Target Gene Details
Entrez Gene ID: | 100289580 |
Gene Name: | uncharacterized LOC100289580 |
Gene Aliases: |
- |
Location: |
Chr.16:88731180-88741425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100289580 | NR_103774.1 | |||
AK127064.1 | 1 | 442 | ||
AK294743.1 |
Target Gene Details
Entrez Gene ID: | 9780 |
Gene Name: | piezo type mechanosensitive ion channel component 1 |
Gene Aliases: |
DHS, FAM38A, LMPH3, Mib |
Location: |
Chr.16:88715338-88785220 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIEZO1 | NM_001142864.3 | NP_001136336.2 | ||
AA682915.1 | ||||
KC602455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1063024 | Chr.16:88707330 - 89238482 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471114 | Chr.16:88688461 - 88756339 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv471113 | Chr.16:88650665 - 88764428 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3064n100 | Chr.16:88683753 - 88846491 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517643 | Chr.16:88713767 - 88756339 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv833327 | Chr.16:88667431 - 88757621 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952070 | Chr.16:88257295 - 88780192 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1927 | Chr.16:88712585 - 88743583 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
nsv1160442 | Chr.16:88708161 - 88778524 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv3065n100 | Chr.16:88711587 - 88927172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)