Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | LOC100505942 ELMO3 PSMB10 PLEKHG4 DPEP3 FBXL8 PSKH1 ENKD1 ACD EXOC3L1 NOL3 TRADD CTCF ATP6V0D1 KIAA0895L EDC4 MIR328 FAM65A C16orf86 E2F4 PARD6A SLC12A4 LOC100131303 GFOD2 HSD11B2 ZDHHC1 SLC9A5 HSF4 DPEP2 LRRC29 CENPT KCTD19 LOC105369155 TMEM208 LRRC36 CTRL THAP11 AGRP LCAT NRN1L DUS2 FHOD1 NUTF2 TPPP3 TSNAXIP1 DDX28 RANBP10 LOC107984813 CARMIL2 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | MIR328 ELMO3 E2F4 B3GNT9 PLEKHG4 FBXL8 FHOD1 LOC105369155 TMEM208 EXOC3L1 SLC9A5 HSF4 NOL3 TRADD C16orf70 KIAA0895L LRRC29 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | MIR328 ELMO3 E2F4 B3GNT9 FBXL8 FHOD1 LOC105369155 TMEM208 EXOC3L1 HSF4 NOL3 TRADD C16orf70 KIAA0895L LRRC29 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | MIR328 ELMO3 EXOC3L1 HSF4 NOL3 E2F4 KIAA0895L LRRC29 FBXL8 LOC105369155 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | MIR328 ELMO3 EXOC3L1 SLC9A5 E2F4 KIAA0895L PLEKHG4 LRRC29 FHOD1 LOC105369155 TMEM208 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | MIR328 ELMO3 E2F4 PLEKHG4 KCTD19 FHOD1 LOC105369155 TMEM208 EXOC3L1 SLC9A5 NOL3 KIAA0895L LRRC29 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | LOC100505942 ELMO3 PSMB10 PLEKHG4 DPEP3 FBXL8 PSKH1 ENKD1 ACD EXOC3L1 NOL3 TRADD CTCF ATP6V0D1 KIAA0895L EDC4 MIR328 FAM65A C16orf86 E2F4 PARD6A SLC12A4 LOC100131303 GFOD2 HSD11B2 ZDHHC1 SLC9A5 HSF4 DPEP2 LRRC29 CENPT KCTD19 LOC105369155 TMEM208 LRRC36 CTRL THAP11 C16orf70 AGRP LCAT B3GNT9 NRN1L DUS2 FHOD1 NUTF2 TPPP3 TSNAXIP1 DDX28 RANBP10 LOC107984813 CARMIL2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |